Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01493547_m1
          See other HNF4G GE Assays ›
          Gene Symbol:
          Hnf4g
          Gene Name:
          hepatocyte nuclear factor 4, gamma
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 100225880 - 100372728 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01493547_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hnf4g
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108939.1 NP_001102409.1 4 - 5 603 96
          XM_006232165.2 XP_006232227.1 4 - 5 1354 96
          XM_006232166.2 XP_006232228.1 5 - 6 1197 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003314.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Nuclear Receptor Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          intracellular receptor signaling pathway
          steroid hormone mediated signaling pathway
          positive regulation of transcription, DNA-templated
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          zinc ion binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P046577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153444 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37209 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86705 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96974 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61414 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135566 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130511 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156055 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101555 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1939 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126279 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121026 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18046 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129757 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118200 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112061 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89504 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P046577 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P046577 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P046577 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P433676 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65645 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5492 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64177 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80243 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11675 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59420 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27468 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10822416 - -
          Affymetrix Rat Gene 1.0 ST Array 10822409 - -
          Affymetrix Rat Gene 1.0 ST Array 10822415 - -
          Affymetrix Rat Gene 1.0 ST Array 10822412 - -
          Affymetrix Rat Gene 1.0 ST Array 10822413 - -
          Affymetrix Rat Gene 1.0 ST Array 10822414 - -
          Affymetrix Rat Gene 1.0 ST Array 10822411 - -
          Affymetrix Rat Gene 1.0 ST Array 10822410 - -
          Affymetrix Rat Gene 1.1 ST Array 10822408 - -
          Affymetrix Rat Clariom S Assay PSR0200022740.rn.1 TC0200003314.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200022739.rn.1 TC0200003314.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10822408 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21354209 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline