Search
Search
| Gene Symbol: Mtfr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100977.1 | NP_001094447.1 | 2 - 3 | 183 | 73 | |
| XM_006232178.2 | XP_006232240.1 | 3 - 4 | 340 | 73 | ||
| XM_006232179.2 | XP_006232241.1 | 3 - 4 | 319 | 73 | ||
| XM_006232180.2 | XP_006232242.1 | 2 - 3 | 1129 | 73 | ||
| GenBank mRNA | EF221637.1 | - | 2 - 3 | 183 | 73 | |
| FQ217237.1 | - | 3 - 4 | 291 | 73 | ||
Set Membership: |
|