Search
Search
| Gene Symbol: Car13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134993.1 | NP_001128465.1 | 2 - 3 | 295 | 76 | |
| GenBank mRNA | BC169052.1 | - | 2 - 3 | 259 | 7386405 | 76 |
| FQ220871.1 | - | 2 - 3 | 257 | 76 | ||
Set Membership: |
|