Search
Search
| Gene Symbol: Nim1k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006231982.2 | XP_006232044.1 | 3 - 4 | 3029 | 99 | |
| XM_008760801.1 | XP_008759023.1 | 3 - 4 | 1543 | 99 | ||
| XM_227081.6 | XP_227081.3 | 3 - 4 | 2159 | 99 | ||
Set Membership: |
|