Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01493959_m1
          See other HMGCS1 GE Assays ›
          Gene Symbol:
          Hmgcs1
          Gene Name:
          3-hydroxy-3-methylglutaryl-CoA synthase 1 (soluble)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 52427351 - 52445082 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01493959_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hmgcs1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017268.1 NP_058964.1 7 - 8 1170 80
          XM_006231959.1 XP_006232021.1 6 - 7 1339 80
          GenBank mRNA AY724522.1 - 6 - 7 1156 80
          FQ211444.1 - 6 - 7 1142 80
          X52625.1 - 7 - 8 1170 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000469.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hmgcs1

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          cholesterol biosynthetic process
          brain development
          isoprenoid biosynthetic process
          male gonad development
          response to low light intensity stimulus
          response to hormone
          response to organic substance
          response to organonitrogen compound
          response to organic cyclic compound
          response to purine-containing compound
          response to vitamin E
          response to gonadotropin
          response to drug
          response to tellurium ion
          response to lipoprotein particle
          response to cholesterol
          cellular response to follicle-stimulating hormone stimulus
          cellular response to cholesterol
          cellular response to organic cyclic compound
          hydroxymethylglutaryl-CoA synthase activity
          drug binding
          isomerase activity
          protein homodimerization activity
          organic acid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P151071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287435 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132326 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108632 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P151071 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P151071 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P151071 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P487240 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11890 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700147478 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57387 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61075 - -
          Agilent Rat cDNA Microarray (G4105A) 700147478 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10813220 - -
          Affymetrix Rat Gene 1.0 ST Array 10813216 - -
          Affymetrix Rat Gene 1.0 ST Array 10813217 - -
          Affymetrix Rat Gene 1.0 ST Array 10813223 - -
          Affymetrix Rat Gene 1.0 ST Array 10813218 - -
          Affymetrix Rat Gene 1.0 ST Array 10813222 - -
          Affymetrix Rat Gene 1.0 ST Array 10813219 - -
          Affymetrix Rat Gene 1.0 ST Array 10813221 - -
          Affymetrix Rat Gene 1.0 ST Array 10813215 - -
          Affymetrix Rat Gene 1.0 ST Array 10813224 - -
          Affymetrix Rat Gene 1.1 ST Array 10813214 - -
          Rat Genome 230 2.0 Array 1367932_at - -
          Rat Genome 230 2.0 Array 1367932_PM_at - -
          Affymetrix HT Rat Focus Array 1367932_at - -
          Affymetrix HT Rat Focus Array 1367932_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200002712.rn.1 TC0200000469.rn.1 RTA TC
          Rat Expression Set 230 1367932_at - -
          Rat Expression Set 230 1367932_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10813214 - -
          Rat Genome U34 Set X52625_at - -
          Rat Genome U34 Set rc_AI177004_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21847002 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376480 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline