Search
Search
| Gene Symbol: Ccnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC059113.1 | - | 6 - 7 | 6922392 | 82 | |
| EV765751.1 | - | 2 - 3 | 416 | 82 | ||
| FN800627.1 | - | 4 - 5 | 664 | 82 | ||
| FN800629.1 | - | 1 - 2 | 120 | 82 | ||
| L11995.1 | - | 6 - 7 | 82 | |||
| X60768.1 | - | 6 - 7 | 941 | 82 | ||
Set Membership: |
|