Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01494691_m1
          See other SELT GE Assays ›
          Gene Symbol:
          Selt
          Gene Name:
          selenoprotein T
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 148765412 - 148781080 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01494691_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Selt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014253.2 NP_001014275.2 4 - 5 521 92
          GenBank mRNA AY995234.1 - 4 - 5 521 92
          BC086953.1 - 4 - 5 415 7321890 92
          FQ211813.1 - 539 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001209.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Selt

          Gene Ontology Categories:

          Function(s) Process(es)

          response to glucose
          pancreas development
          insulin secretion involved in cellular response to glucose stimulus
          glucose homeostasis
          cell redox homeostasis
          selenium binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P526425 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P110445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171360 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135029 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93327 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P702492 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P526425 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P526425 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P702492 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P702492 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P526425 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P526425 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P338207 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101463 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87842 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10815555 - -
          Affymetrix Rat Gene 1.0 ST Array 10815553 - -
          Affymetrix Rat Gene 1.0 ST Array 10815557 - -
          Affymetrix Rat Gene 1.0 ST Array 10815554 - -
          Affymetrix Rat Gene 1.0 ST Array 10815559 - -
          Affymetrix Rat Gene 1.0 ST Array 10815556 - -
          Affymetrix Rat Gene 1.0 ST Array 10815560 - -
          Affymetrix Rat Gene 1.0 ST Array 10815561 - -
          Affymetrix Rat Gene 1.0 ST Array 10815558 - -
          Affymetrix Rat Gene 1.1 ST Array 10815552 - -
          Rat Genome 230 2.0 Array 1373508_at - -
          Rat Genome 230 2.0 Array 1373508_PM_at - -
          Affymetrix HT Rat Focus Array 1373508_at - -
          Affymetrix HT Rat Focus Array 1373508_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200007658.rn.1 TC0200001209.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200007659.rn.1 TC0200001209.rn.1 RTA TC
          Rat Expression Set 230 1373508_PM_at - -
          Rat Expression Set 230 1373508_at - -
          Affymetrix Rat Gene 1.0 ST Array 10815552 - -
          Rat Genome U34 Set rc_AI169293_at - -
          Rat Genome U34 Set rc_AI180004_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21301803 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline