Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01494726_m1
          See other ERICH6 GE Assays ›
          Gene Symbol:
          Erich6
          Gene Name:
          glutamate-rich 6
          Gene Aliases:
          Fam194a
          Chromosome Location:
          Chr.2: 148812060 - 148838162 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01494726_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Erich6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024300.1 NP_001019471.1 13 - 14 1702 106
          GenBank mRNA BC083834.1 - 13 - 14 1702 7112954 106

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Erich6

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P073609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850505 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8460 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161838 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27677 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P073609 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P073609 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P073609 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P935156 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50600 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55218 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92557 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10823335 - -
          Affymetrix Rat Gene 1.0 ST Array 10823333 - -
          Affymetrix Rat Gene 1.0 ST Array 10823328 - -
          Affymetrix Rat Gene 1.0 ST Array 10823330 - -
          Affymetrix Rat Gene 1.0 ST Array 10823329 - -
          Affymetrix Rat Gene 1.0 ST Array 10823324 - -
          Affymetrix Rat Gene 1.0 ST Array 10823326 - -
          Affymetrix Rat Gene 1.0 ST Array 10823334 - -
          Affymetrix Rat Gene 1.0 ST Array 10823331 - -
          Affymetrix Rat Gene 1.0 ST Array 10823327 - -
          Affymetrix Rat Gene 1.0 ST Array 10823336 - -
          Affymetrix Rat Gene 1.0 ST Array 10823323 - -
          Affymetrix Rat Gene 1.0 ST Array 10823325 - -
          Affymetrix Rat Gene 1.0 ST Array 10823332 - -
          Affymetrix Rat Gene 1.1 ST Array 10823322 - -
          Affymetrix Rat Gene 1.0 ST Array 10823322 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21775716 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline