Search
Search
| Gene Symbol: Rnf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109444.2 | NP_001102914.1 | 280 | 85 | ||
| GenBank mRNA | BC081881.1 | - | 280 | 7108181 | 85 | |
| FQ224754.1 | - | 448 | 85 | |||
Set Membership: |
|