Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01495144_g1
          See other ELF2 GE Assays ›
          Gene Symbol:
          Elf2
          Gene Name:
          E74-like factor 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 140310374 - 140399322 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01495144_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Elf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012181.2 NP_001012181.1 6 - 7 1712 66
          NM_001033909.1 NP_001029081.1 8 - 9 1392 66
          XM_006232325.2 XP_006232387.1 9 - 10 1594 66
          XM_006232327.2 XP_006232389.1 6 - 7 3780 66
          XM_006232328.1 XP_006232390.1 7 - 8 1106 66
          XM_006232329.1 XP_006232391.1 7 - 8 1070 66
          XM_008760992.1 XP_008759214.1 8 - 9 1977 66
          GenBank mRNA BC083585.1 - 9 - 10 1544 7131145 66
          BC083598.1 - 6 - 7 1064 7132941 66

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Elf2

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P730411 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P278938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282448 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52645 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176913 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55568 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19144 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114031 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106130 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142942 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41989 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P278938 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P730411 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P278938 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P730411 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P278938 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P730411 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P278938 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P730411 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P585615 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24782 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3974 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56674 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86762 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4689 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8451 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32512 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10823083 - -
          Affymetrix Rat Gene 1.0 ST Array 10823076 - -
          Affymetrix Rat Gene 1.0 ST Array 10823077 - -
          Affymetrix Rat Gene 1.0 ST Array 10823079 - -
          Affymetrix Rat Gene 1.0 ST Array 10823081 - -
          Affymetrix Rat Gene 1.0 ST Array 10823088 - -
          Affymetrix Rat Gene 1.0 ST Array 10823082 - -
          Affymetrix Rat Gene 1.0 ST Array 10823086 - -
          Affymetrix Rat Gene 1.0 ST Array 10823090 - -
          Affymetrix Rat Gene 1.0 ST Array 10823087 - -
          Affymetrix Rat Gene 1.0 ST Array 10823084 - -
          Affymetrix Rat Gene 1.0 ST Array 10823085 - -
          Affymetrix Rat Gene 1.0 ST Array 10823078 - -
          Affymetrix Rat Gene 1.0 ST Array 10823089 - -
          Affymetrix Rat Gene 1.0 ST Array 10823080 - -
          Affymetrix Rat Gene 1.1 ST Array 10823075 - -
          Rat Genome 230 2.0 Array 1382637_PM_at - -
          Rat Genome 230 2.0 Array 1382637_at - -
          Rat Genome 230 2.0 Array 1389130_at - -
          Rat Genome 230 2.0 Array 1379954_at - -
          Rat Genome 230 2.0 Array 1379954_PM_at - -
          Rat Genome 230 2.0 Array 1389130_PM_at - -
          Affymetrix HT Rat Focus Array 1382637_at - -
          Affymetrix HT Rat Focus Array 1389130_at - -
          Affymetrix HT Rat Focus Array 1382637_PM_at - -
          Affymetrix HT Rat Focus Array 1379954_PM_at - -
          Affymetrix HT Rat Focus Array 1389130_PM_at - -
          Affymetrix HT Rat Focus Array 1379954_at - -
          Affymetrix Rat Clariom S Assay PSR0200024538.rn.1 TC0200003643.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200024535.rn.1 TC0200003643.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200024536.rn.1 TC0200003643.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200024537.rn.1 TC0200003643.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200024539.rn.1 TC0200003643.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200024540.rn.1 TC0200003643.rn.1 RTA TC
          Rat Expression Set 230 1389130_PM_at - -
          Rat Expression Set 230 1389130_at - -
          Rat Expression Set 230 1379954_at - -
          Rat Expression Set 230 1379954_PM_at - -
          Rat Expression Set 230 1382637_at - -
          Rat Expression Set 230 1382637_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10823075 - -
          Rat Genome U34 Set rc_AA899415_at - -
          Rat Genome U34 Set rc_AI137223_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375290 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376700 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-fcf5f9d6d-dcxfx:80/100.66.77.77:80.
          git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.51.0-2026.05.21-1.0