Search
Search
| Gene Symbol: Map4k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133407.1 | NP_596898.1 | 27 - 28 | 2208 | 64 | |
| XM_006239656.2 | XP_006239718.1 | 28 - 29 | 2395 | 64 | ||
| GenBank mRNA | AF312224.1 | - | 27 - 28 | 2040 | 64 | |
Set Membership: |
|