Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01495391_m1
          See other ASH1L GE Assays ›
          Gene Symbol:
          Ash1l
          Gene Name:
          ash1 (absent, small, or homeotic)-like (Drosophila)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 188252645 - 188389251 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01495391_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ash1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107689.1 NP_001101159.1 15 - 16 7492 70
          XM_008761153.1 XP_008759375.1 17 - 18 7587 70
          XM_008761154.1 XP_008759376.1 16 - 17 7455 70
          XM_008761155.1 XP_008759377.1 17 - 18 7739 70
          XM_008761156.1 XP_008759378.1 18 - 19 7874 70
          XM_008761157.1 XP_008759379.1 18 - 19 7760 70
          XM_008761158.1 XP_008759380.1 17 - 18 7656 70
          XM_008761159.1 XP_008759381.1 17 - 18 7504 70
          XM_008761160.1 XP_008759382.1 16 - 17 7455 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001560.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of acute inflammatory response
          peptidyl-lysine methylation
          interleukin-6 production
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of MAPK cascade
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of inflammatory response
          histone H3-K4 methylation
          DNA binding
          chromatin binding
          zinc ion binding
          histone methyltransferase activity (H3-K4 specific)

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P165466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16453 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120868 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126534 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125014 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141902 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125217 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52136 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139784 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5601 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26794 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58464 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153444 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44331 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132886 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6042 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77872 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86606 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30112 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P165466 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P165466 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P165466 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P165466 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P389283 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P434386 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P879104 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92531 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84942 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24013 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74552 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10816651 - -
          Affymetrix Rat Gene 1.0 ST Array 10816669 - -
          Affymetrix Rat Gene 1.0 ST Array 10816655 - -
          Affymetrix Rat Gene 1.0 ST Array 10816658 - -
          Affymetrix Rat Gene 1.0 ST Array 10816664 - -
          Affymetrix Rat Gene 1.0 ST Array 10816667 - -
          Affymetrix Rat Gene 1.0 ST Array 10816652 - -
          Affymetrix Rat Gene 1.0 ST Array 10816663 - -
          Affymetrix Rat Gene 1.0 ST Array 10816661 - -
          Affymetrix Rat Gene 1.0 ST Array 10816654 - -
          Affymetrix Rat Gene 1.0 ST Array 10816659 - -
          Affymetrix Rat Gene 1.0 ST Array 10816665 - -
          Affymetrix Rat Gene 1.0 ST Array 10816673 - -
          Affymetrix Rat Gene 1.0 ST Array 10816660 - -
          Affymetrix Rat Gene 1.0 ST Array 10816668 - -
          Affymetrix Rat Gene 1.0 ST Array 10816662 - -
          Affymetrix Rat Gene 1.0 ST Array 10816671 - -
          Affymetrix Rat Gene 1.0 ST Array 10816650 - -
          Affymetrix Rat Gene 1.0 ST Array 10816674 - -
          Affymetrix Rat Gene 1.0 ST Array 10816672 - -
          Affymetrix Rat Gene 1.0 ST Array 10816670 - -
          Affymetrix Rat Gene 1.0 ST Array 10816666 - -
          Affymetrix Rat Gene 1.0 ST Array 10816653 - -
          Affymetrix Rat Gene 1.1 ST Array 10816649 - -
          Rat Genome 230 2.0 Array 1382847_PM_at - -
          Rat Genome 230 2.0 Array 1382862_PM_at - -
          Rat Genome 230 2.0 Array 1381291_at - -
          Rat Genome 230 2.0 Array 1380497_PM_at - -
          Rat Genome 230 2.0 Array 1382847_at - -
          Rat Genome 230 2.0 Array 1381291_PM_at - -
          Rat Genome 230 2.0 Array 1372785_PM_at - -
          Rat Genome 230 2.0 Array 1382862_at - -
          Rat Genome 230 2.0 Array 1380497_at - -
          Rat Genome 230 2.0 Array 1372785_at - -
          Affymetrix HT Rat Focus Array 1372785_at - -
          Affymetrix HT Rat Focus Array 1372785_PM_at - -
          Affymetrix HT Rat Focus Array 1382847_at - -
          Affymetrix HT Rat Focus Array 1380497_at - -
          Affymetrix HT Rat Focus Array 1382847_PM_at - -
          Affymetrix HT Rat Focus Array 1380497_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200010032.rn.1 TC0200001560.rn.1 RTA TC
          Rat Expression Set 230 1372785_PM_at - -
          Rat Expression Set 230 1372785_at - -
          Rat Expression Set 230 1381291_at - -
          Rat Expression Set 230 1382847_PM_at - -
          Rat Expression Set 230 1382862_PM_at - -
          Rat Expression Set 230 1380497_at - -
          Rat Expression Set 230 1382862_at - -
          Rat Expression Set 230 1380497_PM_at - -
          Rat Expression Set 230 1382847_at - -
          Rat Expression Set 230 1381291_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10816649 - -
          Rat Genome U34 Set rc_AI112511_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22321457 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369891 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline