Search
Search

Pip4k2b| Gene Symbol: Pip4k2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053550.1 | NP_446002.1 | 3 - 4 | 473 | 78 | |
| XM_008768131.1 | XP_008766353.1 | 3 - 4 | 752 | 78 | ||
| GenBank mRNA | AF033355.1 | - | 3 - 4 | 473 | 78 | |
Set Membership: |
|