Search
Search
| Gene Symbol: Adprm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009246.1 | NP_001009246.1 | 3 - 4 | 756 | 132 | |
| XM_006246603.2 | XP_006246665.1 | 2 - 3 | 716 | 132 | ||
| XM_006246604.2 | XP_006246666.1 | 3 - 4 | 823 | 132 | ||
| GenBank mRNA | BC088174.1 | - | 3 - 4 | 756 | 7323419 | 132 |
| EU037900.1 | - | 1 - 2 | 603 | 132 | ||
Set Membership: |
|