Search
Search

Rapgef6| Gene Symbol: Rapgef6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107003.1 | NP_001100473.1 | 15 - 16 | 2007 | 80 | |
| XM_006246277.2 | XP_006246339.1 | 15 - 16 | 2056 | 80 | ||
| XM_006246278.2 | XP_006246340.1 | 15 - 16 | 2055 | 80 | ||
| XM_006246279.2 | XP_006246341.1 | 15 - 16 | 2055 | 80 | ||
| XM_006246280.2 | XP_006246342.1 | 15 - 16 | 2055 | 80 | ||
Set Membership: |
|