Search
Search
| Gene Symbol: Ttc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005529.1 | NP_001005529.1 | 5 - 6 | 632 | 66 | |
| XM_008767639.1 | XP_008765861.1 | 5 - 6 | 683 | 66 | ||
| GenBank mRNA | BC082093.1 | - | 5 - 6 | 632 | 7134129 | 66 |
| FQ213119.1 | - | 5 - 6 | 649 | 66 | ||
Set Membership: |
|