Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01496034_m1
          See other WSB1 GE Assays ›
          Gene Symbol:
          Wsb1
          Gene Name:
          WD repeat and SOCS box-containing 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 66586905 - 66602984 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01496034_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wsb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025664.1 NP_001020835.2 3 - 4 790 133
          NM_001042561.1 NP_001036026.1 8 - 9 1202 133
          XM_008768016.1 XP_008766238.1 8 - 9 2125 133
          GenBank mRNA BC098720.1 - 3 - 4 790 133
          BC167746.1 - 8 - 9 1205 8364583 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003359.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Wsb1

          Gene Ontology Categories:

          Function(s)

          protein ubiquitination
          intracellular signal transduction

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P158940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869971 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53745 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47895 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158940 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158940 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P158940 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P274815 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54643 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10745333 - -
          Affymetrix Rat Gene 1.0 ST Array 10745327 - -
          Affymetrix Rat Gene 1.0 ST Array 10745331 - -
          Affymetrix Rat Gene 1.0 ST Array 10745324 - -
          Affymetrix Rat Gene 1.0 ST Array 10745326 - -
          Affymetrix Rat Gene 1.0 ST Array 10745330 - -
          Affymetrix Rat Gene 1.0 ST Array 10745334 - -
          Affymetrix Rat Gene 1.0 ST Array 10745325 - -
          Affymetrix Rat Gene 1.0 ST Array 10745328 - -
          Affymetrix Rat Gene 1.0 ST Array 10745332 - -
          Affymetrix Rat Gene 1.0 ST Array 10745329 - -
          Affymetrix Rat Gene 1.1 ST Array 10745323 - -
          Rat Genome 230 2.0 Array 1374204_at - -
          Rat Genome 230 2.0 Array 1374204_PM_at - -
          Affymetrix HT Rat Focus Array 1374204_PM_at - -
          Affymetrix HT Rat Focus Array 1374204_at - -
          Affymetrix Rat Clariom S Assay PSR1000030093.rn.1 TC1000003359.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000030095.rn.1 TC1000003359.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000030094.rn.1 TC1000003359.rn.1 RTA TC
          Rat Expression Set 230 1374204_at - -
          Rat Expression Set 230 1374204_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10745323 - -
          Rat Genome U34 Set rc_AA963983_at - -
          Rat Genome U34 Set rc_AI176125_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20865222 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357658 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline