Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01496470_m1
          See other DHX40 GE Assays ›
          Gene Symbol:
          Dhx40
          Gene Name:
          DEAH (Asp-Glu-Ala-His) box polypeptide 40
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 74081442 - 74119032 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01496470_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dhx40
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005873.1 NP_001005873.1 17 - 18 2293 121
          GenBank mRNA BC083821.1 - 17 - 18 2293 7111216 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003473.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dhx40

          Panther Classification:

          Molecular Function -

          RNA helicase

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA processing
          ATP-dependent RNA helicase activity
          ATP binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1055584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268057 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67717 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141281 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26426 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1055584 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1055584 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1055584 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1055584 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86977 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55863 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10745975 - -
          Affymetrix Rat Gene 1.0 ST Array 10745980 - -
          Affymetrix Rat Gene 1.0 ST Array 10745985 - -
          Affymetrix Rat Gene 1.0 ST Array 10745991 - -
          Affymetrix Rat Gene 1.0 ST Array 10745992 - -
          Affymetrix Rat Gene 1.0 ST Array 10745986 - -
          Affymetrix Rat Gene 1.0 ST Array 10745979 - -
          Affymetrix Rat Gene 1.0 ST Array 10745987 - -
          Affymetrix Rat Gene 1.0 ST Array 10745982 - -
          Affymetrix Rat Gene 1.0 ST Array 10745981 - -
          Affymetrix Rat Gene 1.0 ST Array 10745976 - -
          Affymetrix Rat Gene 1.0 ST Array 10745984 - -
          Affymetrix Rat Gene 1.0 ST Array 10745983 - -
          Affymetrix Rat Gene 1.0 ST Array 10745977 - -
          Affymetrix Rat Gene 1.0 ST Array 10745988 - -
          Affymetrix Rat Gene 1.0 ST Array 10745993 - -
          Affymetrix Rat Gene 1.0 ST Array 10745990 - -
          Affymetrix Rat Gene 1.0 ST Array 10745978 - -
          Affymetrix Rat Gene 1.0 ST Array 10745989 - -
          Affymetrix Rat Gene 1.1 ST Array 10745974 - -
          Rat Genome 230 2.0 Array 1390545_at - -
          Rat Genome 230 2.0 Array 1390545_PM_at - -
          Rat Genome 230 2.0 Array 1377785_at - -
          Rat Genome 230 2.0 Array 1377785_PM_at - -
          Affymetrix HT Rat Focus Array 1377785_PM_at - -
          Affymetrix HT Rat Focus Array 1377785_at - -
          Affymetrix Rat Clariom S Assay PSR1000031169.rn.1 TC1000003473.rn.1 RTA TC
          Rat Expression Set 230 1377785_PM_at - -
          Rat Expression Set 230 1377785_at - -
          Affymetrix Rat Gene 1.0 ST Array 10745974 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22286436 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365095 - -
          RatRef-12 Expression BeadChip ILMN_66791 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline