Search
Search
| Gene Symbol: Heatr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079897.1 | NP_001073366.1 | 14 - 15 | 2502 | 59 | |
| GenBank mRNA | EF122002.1 | - | 14 - 15 | 2502 | 59 | |
| EF122003.1 | - | 14 - 15 | 2502 | 59 | ||
Set Membership: |
|