Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01496517_g1
          See other CLDN7 GE Assays ›
          Gene Symbol:
          Cldn7
          Gene Name:
          claudin 7
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 56576322 - 56578632 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01496517_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cldn7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031702.1 NP_113890.1 3 - 4 690 67
          XM_006246805.2 XP_006246867.1 2 - 3 890 67
          GenBank mRNA AJ011811.1 - 2 - 3 326 67
          BC101892.1 - 3 - 4 690 7930797 67
          BC159404.1 - 2 - 3 784 7458165 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000938.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cldn7

          Panther Classification:

          Molecular Function -

          tight junction

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cell adhesion
          positive regulation of cell proliferation
          calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules
          negative regulation of protein homooligomerization
          negative regulation of apoptotic process
          positive regulation of cell motility
          structural molecule activity
          identical protein binding
          cell adhesion molecule binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1010902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392573 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1010902 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1010902 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1010902 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1010902 - -
          Agilent Rat cDNA Microarray (G4105A) 700175676 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10735092 - -
          Affymetrix Rat Gene 1.0 ST Array 10735095 - -
          Affymetrix Rat Gene 1.0 ST Array 10735094 - -
          Affymetrix Rat Gene 1.0 ST Array 10735093 - -
          Affymetrix Rat Gene 1.1 ST Array 10735091 - -
          Rat Genome 230 2.0 Array 1370894_at - -
          Rat Genome 230 2.0 Array 1370894_PM_at - -
          Affymetrix HT Rat Focus Array 1370894_PM_at - -
          Affymetrix HT Rat Focus Array 1370894_at - -
          Affymetrix Rat Clariom S Assay PSR1000008216.rn.1 TC1000000938.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008218.rn.1 TC1000000938.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008221.rn.1 TC1000000938.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008222.rn.1 TC1000000938.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008220.rn.1 TC1000000938.rn.1 RTA TC
          Rat Expression Set 230 1370894_at - -
          Rat Expression Set 230 1370894_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10735091 - -
          Rat Genome U34 Set AJ011811_g_at - -
          Rat Genome U34 Set AJ011811_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21855470 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-vppth:80/100.66.77.159:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline