Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01496569_m1
          See other HEXIM2 GE Assays ›
          Gene Symbol:
          Hexim2
          Gene Name:
          hexamethylene bis-acetamide inducible 2
          Gene Aliases:
          RGD1311635
          Chromosome Location:
          Chr.10: 91212290 - 91222841 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01496569_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hexim2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107054.2 NP_001100524.1 4 - 5 447 143
          XM_006247514.2 XP_006247576.1 3 - 4 5092 143
          XM_006247515.2 XP_006247577.1 3 - 4 5095 143
          XM_006247516.2 XP_006247578.1 2 - 3 424 143
          XM_006247517.2 XP_006247579.1 2 - 3 427 143
          XM_006247518.2 XP_006247580.1 2 - 3 5005 143
          XM_006247520.2 XP_006247582.1 2 - 3 330 143
          XM_006247521.2 XP_006247583.1 2 - 3 389 143
          XM_006247522.2 XP_006247584.1 2 - 3 333 143
          XM_008768288.1 XP_008766510.1 2 - 3 925 143
          XM_008768289.1 XP_008766511.1 2 - 3 392 143
          GenBank mRNA BC167046.1 - 4 - 5 447 7132762 143

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001714.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          biological_process
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          negative regulation of transcription, DNA-templated
          molecular_function
          cyclin-dependent protein serine/threonine kinase inhibitor activity
          snRNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P541692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477909 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P541692 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P541692 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P541692 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P541692 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10738675 - -
          Affymetrix Rat Gene 1.0 ST Array 10738674 - -
          Affymetrix Rat Gene 1.1 ST Array 10738673 - -
          Rat Genome 230 2.0 Array 1374732_PM_at - -
          Rat Genome 230 2.0 Array 1374732_at - -
          Affymetrix HT Rat Focus Array 1374732_at - -
          Affymetrix HT Rat Focus Array 1374732_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000015539.rn.1 TC1000001714.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000015554.rn.1 TC1000001714.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000015556.rn.1 TC1000001714.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000015559.rn.1 TC1000001714.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000015541.rn.1 TC1000001714.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000015560.rn.1 TC1000001714.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000015549.rn.1 TC1000001714.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000015550.rn.1 TC1000001714.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000015558.rn.1 TC1000001714.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000015555.rn.1 TC1000001714.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000015557.rn.1 TC1000001714.rn.1 RTA TC
          Rat Expression Set 230 1374732_at - -
          Rat Expression Set 230 1374732_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10738673 - -
          Rat Genome U34 Set rc_AA925492_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650330 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline