Search
Search
| Gene Symbol: Ppy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012626.2 | NP_036758.1 | 2 - 3 | 239 | 67 | |
| GenBank mRNA | BP484139.1 | - | 2 - 3 | 222 | 67 | |
| M13588.1 | - | 2 - 3 | 239 | 67 | ||
Set Membership: |
|