Search
Search
| Gene Symbol: Supt6h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191820.1 | NP_001178749.1 | 28 - 29 | 4192 | 67 | |
| XM_008768006.1 | XP_008766228.1 | 28 - 29 | 3853 | 67 | ||
| XM_008768007.1 | XP_008766229.1 | 29 - 30 | 4044 | 67 | ||
| XM_008768008.1 | XP_008766230.1 | 28 - 29 | 4044 | 67 | ||
Set Membership: |
|