Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tvp23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107568.3 | NP_001101038.2 | 5 - 6 | 499 | 65 | |
GenBank mRNA | BC168178.1 | - | 5 - 6 | 497 | 65 | |
CK483802.1 | - | 5 - 6 | 499 | 65 | ||
FQ212667.1 | - | 5 - 6 | 535 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |