Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01497155_m1
          See other TMEM204 GE Assays ›
          Gene Symbol:
          Tmem204
          Gene Name:
          transmembrane protein 204
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 14417608 - 14443010 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01497155_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem204
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009620.1 NP_001009620.1 2 - 3 993 105
          GenBank mRNA BC087601.1 - 2 - 3 993 7314703 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002414.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem204

          Gene Ontology Categories:

          Function(s)

          lymph vessel development
          regulation of vascular endothelial growth factor receptor signaling pathway
          smooth muscle cell differentiation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P508085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257593 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79434 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34072 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51198 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P508085 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P508085 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P508085 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P508085 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38132 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76115 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10741333 - -
          Affymetrix Rat Gene 1.0 ST Array 10741332 - -
          Affymetrix Rat Gene 1.0 ST Array 10741331 - -
          Affymetrix Rat Gene 1.1 ST Array 10741330 - -
          Rat Genome 230 2.0 Array 1376623_at - -
          Rat Genome 230 2.0 Array 1376623_PM_at - -
          Affymetrix HT Rat Focus Array 1376623_PM_at - -
          Affymetrix HT Rat Focus Array 1376623_at - -
          Affymetrix Rat Clariom S Assay PSR1000021640.rn.1 TC1000002414.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000021642.rn.1 TC1000002414.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000021641.rn.1 TC1000002414.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000021645.rn.1 TC1000002414.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000021644.rn.1 TC1000002414.rn.1 RTA TC
          Rat Expression Set 230 1376623_at - -
          Rat Expression Set 230 1376623_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10741330 - -
          Rat Genome U34 Set rc_AA819523_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21092736 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367558 - -
          RatRef-12 Expression BeadChip ILMN_53038 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline