Search
Search
| Gene Symbol: Ptcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107648.1 | NP_001101118.1 | 3 - 4 | 355 | 67 | |
| XM_006231847.2 | XP_006231909.1 | 3 - 4 | 387 | 67 | ||
| XM_008760675.1 | XP_008758897.1 | 2 - 3 | 1577 | 67 | ||
Set Membership: |
|