Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01498090_g1
          See other LOC499339 GE Assays ›
          Gene Symbol:
          LOC499339
          Gene Name:
          hypothetical protein LOC499339
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 245471939 - 245517076 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01498090_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC499339
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025036.1 NP_001020207.1 16 - 17 1757 109
          XM_006231246.2 XP_006231308.1 16 - 17 1804 109
          GenBank mRNA BC092579.1 - 16 - 17 1757 7323921 109
          BC098744.1 - 15 - 16 1731 7387202 109
          FQ234057.1 - 6 - 7 602 109

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC499339

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of translation
          mRNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P312205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345166 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37775 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43635 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168336 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38605 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154189 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P312205 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P312205 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P312205 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P405655 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1020134 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36690 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71945 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14405 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10729517 - -
          Affymetrix Rat Gene 1.0 ST Array 10729522 - -
          Affymetrix Rat Gene 1.0 ST Array 10940069 - -
          Affymetrix Rat Gene 1.0 ST Array 10729518 - -
          Affymetrix Rat Gene 1.0 ST Array 10729513 - -
          Affymetrix Rat Gene 1.0 ST Array 10940070 - -
          Affymetrix Rat Gene 1.0 ST Array 10729520 - -
          Affymetrix Rat Gene 1.0 ST Array 10729519 - -
          Affymetrix Rat Gene 1.0 ST Array 10729511 - -
          Affymetrix Rat Gene 1.0 ST Array 10729512 - -
          Affymetrix Rat Gene 1.0 ST Array 10729515 - -
          Affymetrix Rat Gene 1.0 ST Array 10729508 - -
          Affymetrix Rat Gene 1.0 ST Array 10940071 - -
          Affymetrix Rat Gene 1.0 ST Array 10729524 - -
          Affymetrix Rat Gene 1.0 ST Array 10729523 - -
          Affymetrix Rat Gene 1.0 ST Array 10729516 - -
          Affymetrix Rat Gene 1.0 ST Array 10729521 - -
          Affymetrix Rat Gene 1.0 ST Array 10729510 - -
          Affymetrix Rat Gene 1.0 ST Array 10729514 - -
          Affymetrix Rat Gene 1.0 ST Array 10729509 - -
          Affymetrix Rat Gene 1.1 ST Array 10940068 - -
          Affymetrix Rat Gene 1.1 ST Array 10729507 - -
          Rat Genome 230 2.0 Array 1376408_at - -
          Rat Genome 230 2.0 Array 1377708_PM_at - -
          Rat Genome 230 2.0 Array 1377708_at - -
          Rat Genome 230 2.0 Array 1376408_PM_at - -
          Rat Genome 230 2.0 Array 1385020_PM_at - -
          Rat Genome 230 2.0 Array 1385020_at - -
          Affymetrix HT Rat Focus Array 1385020_PM_at - -
          Affymetrix HT Rat Focus Array 1376408_PM_at - -
          Affymetrix HT Rat Focus Array 1376408_at - -
          Affymetrix HT Rat Focus Array 1385020_at - -
          Affymetrix HT Rat Focus Array 1377708_PM_at - -
          Affymetrix HT Rat Focus Array 1377708_at - -
          Affymetrix Rat Clariom S Assay PSR0100060292.rn.1 TC0100007486.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100060288.rn.1 TC0100007486.rn.1 RTA TC
          Rat Expression Set 230 1376408_at - -
          Rat Expression Set 230 1376408_PM_at - -
          Rat Expression Set 230 1377708_at - -
          Rat Expression Set 230 1385020_PM_at - -
          Rat Expression Set 230 1377708_PM_at - -
          Rat Expression Set 230 1385020_at - -
          Affymetrix Rat Gene 1.0 ST Array 10940068 - -
          Affymetrix Rat Gene 1.0 ST Array 10729507 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21592679 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354251 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline