Search
Search
| Gene Symbol: Vldlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013155.2 | NP_037287.2 | 4 - 5 | 633 | 76 | |
| XM_006231197.2 | XP_006231259.1 | 5 - 6 | 938 | 76 | ||
| XM_006231198.2 | XP_006231260.1 | 5 - 6 | 667 | 76 | ||
| XM_006231199.2 | XP_006231261.1 | 4 - 5 | 541 | 76 | ||
| GenBank mRNA | L35767.1 | - | 4 - 5 | 633 | 76 | |
Set Membership: |
|