Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01498533_m1
          See other TMEM143 GE Assays ›
          Gene Symbol:
          Tmem143
          Gene Name:
          transmembrane protein 143
          Gene Aliases:
          RGD1305013
          Chromosome Location:
          Chr.1: 101884019 - 101900070 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01498533_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem143
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107513.1 NP_001100983.1 4 - 5 657 133
          XM_006229089.2 XP_006229151.1 4 - 5 652 133
          XM_006229090.2 XP_006229152.1 3 - 4 518 133
          XM_006229092.2 XP_006229154.1 4 - 5 623 133
          XM_008759386.1 XP_008757608.1 4 - 5 623 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001518.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          hematopoietic progenitor cell differentiation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P121608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814819 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8983 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79623 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130729 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P121608 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P121608 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P121608 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P245741 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83895 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47078 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103258 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10706998 - -
          Affymetrix Rat Gene 1.0 ST Array 10706999 - -
          Affymetrix Rat Gene 1.0 ST Array 10706997 - -
          Affymetrix Rat Gene 1.0 ST Array 10707000 - -
          Affymetrix Rat Gene 1.0 ST Array 10707001 - -
          Affymetrix Rat Gene 1.0 ST Array 10706995 - -
          Affymetrix Rat Gene 1.0 ST Array 10706996 - -
          Affymetrix Rat Gene 1.0 ST Array 10707002 - -
          Affymetrix Rat Gene 1.1 ST Array 10706994 - -
          Rat Genome 230 2.0 Array 1382388_PM_at - -
          Rat Genome 230 2.0 Array 1382388_at - -
          Affymetrix HT Rat Focus Array 1382388_PM_at - -
          Affymetrix HT Rat Focus Array 1382388_at - -
          Affymetrix Rat Clariom S Assay PSR0100012149.rn.1 TC0100001518.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012151.rn.1 TC0100001518.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012157.rn.1 TC0100001518.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012150.rn.1 TC0100001518.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012152.rn.1 TC0100001518.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012148.rn.1 TC0100001518.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012161.rn.1 TC0100001518.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012158.rn.1 TC0100001518.rn.1 RTA TC
          Rat Expression Set 230 1382388_at - -
          Rat Expression Set 230 1382388_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10706994 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21427246 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1651133 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline