Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01499024_m1
          See other HPCAL1 GE Assays ›
          Gene Symbol:
          Hpcal1
          Gene Name:
          hippocalcin-like 1
          Gene Aliases:
          NVP-3, Nvp3, VILIP-3
          Chromosome Location:
          Chr.6: 43001948 - 43109083 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01499024_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hpcal1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017356.2 NP_059052.1 4 - 5 819 90
          XM_006239914.2 XP_006239976.1 5 - 6 1426 90
          XM_006239916.2 XP_006239978.1 4 - 5 1321 90
          XM_008764605.1 XP_008762827.1 4 - 5 936 90
          XM_008764606.1 XP_008762828.1 6 - 7 1536 90
          GenBank mRNA BC088759.1 - 4 - 5 819 7317905 90
          D13126.1 - 4 - 5 774 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600000488.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hpcal1

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P231597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408204 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100759 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139421 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4175 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52458 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45510 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53672 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19013 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158461 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66400 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40050 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137074 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32569 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136304 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104525 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62724 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91734 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P231597 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P231597 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P231597 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P231597 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3590 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50977 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50503 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54349 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98142 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97979 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68788 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39795 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10883802 - -
          Affymetrix Rat Gene 1.0 ST Array 10883805 - -
          Affymetrix Rat Gene 1.0 ST Array 10883803 - -
          Affymetrix Rat Gene 1.0 ST Array 10883807 - -
          Affymetrix Rat Gene 1.0 ST Array 10883806 - -
          Affymetrix Rat Gene 1.0 ST Array 10883804 - -
          Affymetrix Rat Gene 1.1 ST Array 10883801 - -
          Rat Genome 230 2.0 Array 1369562_PM_at - -
          Rat Genome 230 2.0 Array 1369562_at - -
          Rat Genome 230 2.0 Array 1372179_at - -
          Rat Genome 230 2.0 Array 1372179_PM_at - -
          Affymetrix HT Rat Focus Array 1372179_at - -
          Affymetrix HT Rat Focus Array 1369562_at - -
          Affymetrix HT Rat Focus Array 1372179_PM_at - -
          Affymetrix HT Rat Focus Array 1369562_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600003347.rn.1 TC0600000488.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600003348.rn.1 TC0600000488.rn.1 RTA TC
          Rat Expression Set 230 1369562_PM_at - -
          Rat Expression Set 230 1372179_at - -
          Rat Expression Set 230 1372179_PM_at - -
          Rat Expression Set 230 1369562_at - -
          Affymetrix Rat Gene 1.0 ST Array 10883801 - -
          Rat Genome U34 Set D13126_at - -
          Rat Genome U34 Set rc_AI101921_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21369027 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline