Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01499073_m1
          See other ABCG5 GE Assays ›
          Gene Symbol:
          Abcg5
          Gene Name:
          ATP-binding cassette, subfamily G (WHITE), member 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 7935771 - 7961339 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01499073_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abcg5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053754.2 NP_446206.2 12 - 13 1829 70
          XM_008764452.1 XP_008762674.1 13 - 14 2069 70
          GenBank mRNA AF312714.3 - 12 - 13 1829 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001823.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Abcg5

          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter primary active transporter transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          response to nutrient
          excretion
          metabolic process
          response to ionizing radiation
          negative regulation of intestinal phytosterol absorption
          sterol transport
          regulation of intestinal cholesterol absorption
          cholesterol transport
          cholesterol efflux
          response to drug
          cholesterol homeostasis
          negative regulation of intestinal cholesterol absorption
          ATP binding
          ATPase activity
          cholesterol transporter activity
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721721 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97967 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1254 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66859 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12924 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12924 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12924 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12924 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38822 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99652 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10888200 - -
          Affymetrix Rat Gene 1.0 ST Array 10888209 - -
          Affymetrix Rat Gene 1.0 ST Array 10888201 - -
          Affymetrix Rat Gene 1.0 ST Array 10888206 - -
          Affymetrix Rat Gene 1.0 ST Array 10888204 - -
          Affymetrix Rat Gene 1.0 ST Array 10888198 - -
          Affymetrix Rat Gene 1.0 ST Array 10888205 - -
          Affymetrix Rat Gene 1.0 ST Array 10888208 - -
          Affymetrix Rat Gene 1.0 ST Array 10888203 - -
          Affymetrix Rat Gene 1.0 ST Array 10888207 - -
          Affymetrix Rat Gene 1.0 ST Array 10888202 - -
          Affymetrix Rat Gene 1.0 ST Array 10888199 - -
          Affymetrix Rat Gene 1.0 ST Array 10888197 - -
          Affymetrix Rat Gene 1.1 ST Array 10888196 - -
          Rat Genome 230 2.0 Array 1369455_PM_at - -
          Rat Genome 230 2.0 Array 1369455_at - -
          Affymetrix HT Rat Focus Array 1369455_PM_at - -
          Affymetrix HT Rat Focus Array 1369455_at - -
          Affymetrix Rat Clariom S Assay PSR0600012941.rn.1 TC0600001823.rn.1 RTA TC
          Rat Expression Set 230 1369455_at - -
          Rat Expression Set 230 1369455_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10888196 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21267604 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358631 - -
          RatRef-12 Expression BeadChip ILMN_53713 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline