Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01499137_m1
          See other RDH11 GE Assays ›
          Gene Symbol:
          Rdh11
          Gene Name:
          retinol dehydrogenase 11 (all-trans/9-cis/11-cis)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 102356498 - 102372618 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01499137_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rdh11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012193.1 NP_001012193.1 6 - 7 931 80
          GenBank mRNA AY387097.1 - 6 - 7 1099 80
          BC079276.1 - 6 - 7 931 7134077 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600002735.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rdh11

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          adaptation of rhodopsin mediated signaling
          retinol metabolic process
          retinal metabolic process
          oxidation-reduction process
          molecular_function
          oxidoreductase activity
          NADP-retinol dehydrogenase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P050545 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P434216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591254 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160261 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143894 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P434216 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P050545 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P434216 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P050545 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P050545 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P434216 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P434216 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35581 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79940 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10890797 - -
          Affymetrix Rat Gene 1.0 ST Array 10890800 - -
          Affymetrix Rat Gene 1.0 ST Array 10890802 - -
          Affymetrix Rat Gene 1.0 ST Array 10890798 - -
          Affymetrix Rat Gene 1.0 ST Array 10890796 - -
          Affymetrix Rat Gene 1.0 ST Array 10890808 - -
          Affymetrix Rat Gene 1.0 ST Array 10890801 - -
          Affymetrix Rat Gene 1.0 ST Array 10890806 - -
          Affymetrix Rat Gene 1.0 ST Array 10890803 - -
          Affymetrix Rat Gene 1.0 ST Array 10890799 - -
          Affymetrix Rat Gene 1.1 ST Array 10890795 - -
          Rat Genome 230 2.0 Array 1373918_PM_at - -
          Rat Genome 230 2.0 Array 1373918_at - -
          Affymetrix HT Rat Focus Array 1373918_at - -
          Affymetrix HT Rat Focus Array 1373918_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600019387.rn.1 TC0600002735.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600019388.rn.1 TC0600002735.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600019386.rn.1 TC0600002735.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600019385.rn.1 TC0600002735.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600019389.rn.1 TC0600002735.rn.1 RTA TC
          Rat Expression Set 230 1373918_PM_at - -
          Rat Expression Set 230 1373918_at - -
          Affymetrix Rat Gene 1.0 ST Array 10890795 - -
          Rat Genome U34 Set rc_AI236036_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21890995 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375818 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376961 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline