Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01499195_m1
          See other DTNB GE Assays ›
          Gene Symbol:
          Dtnb
          Gene Name:
          dystrobrevin, beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 27975305 - 28177236 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01499195_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dtnb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012191.1 NP_001012191.1 18 - 19 3068 55
          XM_006239816.2 XP_006239878.1 18 - 19 3007 55
          XM_006239817.2 XP_006239879.1 20 - 21 2125 55
          XM_006239818.2 XP_006239880.1 19 - 20 2084 55
          XM_006239819.2 XP_006239881.1 19 - 20 2036 55
          XM_006239822.2 XP_006239884.1 18 - 19 2014 55
          GenBank mRNA BC081889.1 - 18 - 19 3068 7109493 55

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600000327.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dtnb

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Process(es)

          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P792760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485756 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61591 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38465 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122360 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64847 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155674 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112763 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64622 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87215 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93513 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32536 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163259 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103819 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11072 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17403 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54976 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101603 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92500 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106664 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88744 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132527 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71614 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38214 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2403 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P792760 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P792760 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P792760 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P347270 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61556 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5137 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30729 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54851 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54848 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86443 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25486 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51459 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93334 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104682 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28404 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45621 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41385 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57156 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13549 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10883337 - -
          Affymetrix Rat Gene 1.0 ST Array 10883351 - -
          Affymetrix Rat Gene 1.0 ST Array 10883353 - -
          Affymetrix Rat Gene 1.0 ST Array 10883350 - -
          Affymetrix Rat Gene 1.0 ST Array 10883352 - -
          Affymetrix Rat Gene 1.0 ST Array 10883349 - -
          Affymetrix Rat Gene 1.0 ST Array 10883346 - -
          Affymetrix Rat Gene 1.0 ST Array 10883341 - -
          Affymetrix Rat Gene 1.0 ST Array 10883356 - -
          Affymetrix Rat Gene 1.0 ST Array 10883340 - -
          Affymetrix Rat Gene 1.0 ST Array 10883338 - -
          Affymetrix Rat Gene 1.0 ST Array 10883342 - -
          Affymetrix Rat Gene 1.0 ST Array 10883348 - -
          Affymetrix Rat Gene 1.0 ST Array 10883345 - -
          Affymetrix Rat Gene 1.0 ST Array 10883347 - -
          Affymetrix Rat Gene 1.0 ST Array 10883344 - -
          Affymetrix Rat Gene 1.0 ST Array 10883343 - -
          Affymetrix Rat Gene 1.0 ST Array 10883339 - -
          Affymetrix Rat Gene 1.0 ST Array 10883336 - -
          Affymetrix Rat Gene 1.0 ST Array 10883355 - -
          Affymetrix Rat Gene 1.1 ST Array 10883335 - -
          Rat Genome 230 2.0 Array 1396409_at - -
          Rat Genome 230 2.0 Array 1376130_PM_a_at - -
          Rat Genome 230 2.0 Array 1376130_a_at - -
          Rat Genome 230 2.0 Array 1396409_PM_at - -
          Affymetrix HT Rat Focus Array 1396409_at - -
          Affymetrix HT Rat Focus Array 1376130_a_at - -
          Affymetrix HT Rat Focus Array 1376130_PM_a_at - -
          Affymetrix HT Rat Focus Array 1396409_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600002269.rn.1 TC0600000327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600002267.rn.1 TC0600000327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600002268.rn.1 TC0600000327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600002270.rn.1 TC0600000327.rn.1 RTA TC
          Rat Expression Set 230 1376130_PM_a_at - -
          Rat Expression Set 230 1376130_a_at - -
          Rat Expression Set 230 1396409_at - -
          Rat Expression Set 230 1396409_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10883335 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21799296 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375274 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376949 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline