Search
Search
| Gene Symbol: Asphd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009716.1 | NP_001009716.1 | 2 - 3 | 1357 | 62 | |
| XM_008769345.1 | XP_008767567.1 | 3 - 4 | 2590 | 62 | ||
| XM_008769346.1 | XP_008767568.1 | 4 - 5 | 2820 | 62 | ||
| XM_008769347.1 | XP_008767569.1 | 4 - 5 | 1780 | 62 | ||
| XM_008769348.1 | XP_008767570.1 | 4 - 5 | 2493 | 62 | ||
| XM_008769349.1 | XP_008767571.1 | 4 - 5 | 2376 | 62 | ||
| XM_008769350.1 | XP_008767572.1 | 3 - 4 | 2278 | 62 | ||
| XM_008769351.1 | XP_008767573.1 | 2 - 3 | 1236 | 62 | ||
| GenBank mRNA | BC088863.1 | - | 2 - 3 | 1357 | 7313227 | 62 |
Set Membership: |
|