Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01499386_m1
          See other UNG GE Assays ›
          Gene Symbol:
          Ung
          Gene Name:
          uracil-DNA glycosylase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 48246593 - 48256157 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01499386_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ung
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013124.1 NP_001013142.1 2 - 3 464 68
          XM_008769336.1 XP_008767558.1 3 - 4 1135 68
          GenBank mRNA BC091211.1 - 2 - 3 464 7381643 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001786.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ung

          Panther Classification:

          Molecular Function -

          DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          base-excision repair
          somatic hypermutation of immunoglobulin genes
          somatic recombination of immunoglobulin gene segments
          negative regulation of apoptotic process
          uracil DNA N-glycosylase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P100443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513240 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79249 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18890 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P100443 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P100443 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P100443 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P100443 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52645 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10762853 - -
          Affymetrix Rat Gene 1.0 ST Array 10762851 - -
          Affymetrix Rat Gene 1.0 ST Array 10762854 - -
          Affymetrix Rat Gene 1.0 ST Array 10762852 - -
          Affymetrix Rat Gene 1.0 ST Array 10762855 - -
          Affymetrix Rat Gene 1.0 ST Array 10762850 - -
          Affymetrix Rat Gene 1.0 ST Array 10762849 - -
          Affymetrix Rat Gene 1.1 ST Array 10762848 - -
          Rat Genome 230 2.0 Array 1389738_PM_at - -
          Rat Genome 230 2.0 Array 1389738_at - -
          Affymetrix HT Rat Focus Array 1389738_at - -
          Affymetrix HT Rat Focus Array 1389738_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200015364.rn.1 TC1200001786.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015363.rn.1 TC1200001786.rn.1 RTA TC
          Rat Expression Set 230 1389738_at - -
          Rat Expression Set 230 1389738_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10762848 - -
          Rat Genome U34 Set AA848421_at - -
          Rat Genome U34 Set rc_AA848420_at - -
          Rat Genome U34 Set AA848421_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21322069 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361057 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline