Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01499500_m1
          See other ANKRD13A GE Assays ›
          Gene Symbol:
          Ankrd13a
          Gene Name:
          ankyrin repeat domain 13a
          Gene Aliases:
          Ankrd13
          Chromosome Location:
          Chr.12: 47558587 - 47588872 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01499500_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ankrd13a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012148.1 NP_001012148.1 14 - 15 1694 71
          XM_006249461.1 XP_006249523.1 14 - 15 2234 71
          XM_006249462.2 XP_006249524.1 14 - 15 1578 71
          GenBank mRNA BC085774.1 - 14 - 15 1694 7096477 71

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ankrd13a

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P499273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155655 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6876 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71409 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43771 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113539 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P499273 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P499273 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P499273 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P235165 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P292836 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11463 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9751 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85516 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10762763 - -
          Affymetrix Rat Gene 1.0 ST Array 10762765 - -
          Affymetrix Rat Gene 1.0 ST Array 10762772 - -
          Affymetrix Rat Gene 1.0 ST Array 10762767 - -
          Affymetrix Rat Gene 1.0 ST Array 10762774 - -
          Affymetrix Rat Gene 1.0 ST Array 10762766 - -
          Affymetrix Rat Gene 1.0 ST Array 10762768 - -
          Affymetrix Rat Gene 1.0 ST Array 10762769 - -
          Affymetrix Rat Gene 1.0 ST Array 10762773 - -
          Affymetrix Rat Gene 1.0 ST Array 10762764 - -
          Affymetrix Rat Gene 1.0 ST Array 10762770 - -
          Affymetrix Rat Gene 1.0 ST Array 10762771 - -
          Affymetrix Rat Gene 1.1 ST Array 10762762 - -
          Rat Genome 230 2.0 Array 1396195_at - -
          Rat Genome 230 2.0 Array 1373059_at - -
          Rat Genome 230 2.0 Array 1373059_PM_at - -
          Rat Genome 230 2.0 Array 1396195_PM_at - -
          Affymetrix HT Rat Focus Array 1396195_PM_at - -
          Affymetrix HT Rat Focus Array 1373059_PM_at - -
          Affymetrix HT Rat Focus Array 1396195_at - -
          Affymetrix HT Rat Focus Array 1373059_at - -
          Affymetrix Rat Clariom S Assay PSR1200015123.rn.1 TC1200001771.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015125.rn.1 TC1200001771.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015122.rn.1 TC1200001771.rn.1 RTA TC
          Rat Expression Set 230 1373059_at - -
          Rat Expression Set 230 1373059_PM_at - -
          Rat Expression Set 230 1396195_at - -
          Rat Expression Set 230 1396195_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10762762 - -
          Rat Genome U34 Set rc_AA944355_at - -
          Rat Genome U34 Set rc_AI178000_at - -
          Rat Genome U34 Set rc_AI231083_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21791792 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376611 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline