Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01499664_m1
          See other ATXN2 GE Assays ›
          Gene Symbol:
          Atxn2
          Gene Name:
          ataxin 2
          Gene Aliases:
          Sca2
          Chromosome Location:
          Chr.12: 40264601 - 40335593 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01499664_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atxn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008769281.1 XP_008767503.1 14 - 15 5209 62
          XM_008769282.1 XP_008767504.1 14 - 15 5209 62
          XM_008769283.1 XP_008767505.1 14 - 15 5208 62
          XM_008769284.1 XP_008767506.1 14 - 15 5208 62
          XM_008769285.1 XP_008767507.1 14 - 15 5208 62
          XM_008769286.1 XP_008767508.1 14 - 15 5208 62
          XM_008769287.1 XP_008767509.1 14 - 15 5209 62
          XM_008769288.1 XP_008767510.1 14 - 15 5208 62
          XM_008769289.1 XP_008767511.1 14 - 15 5209 62
          XM_008769290.1 XP_008767512.1 14 - 15 5208 62
          XM_008769292.1 XP_008767514.1 13 - 14 4999 62
          XM_008769293.1 XP_008767515.1 14 - 15 5208 62
          XM_008769294.1 XP_008767516.1 14 - 15 5209 62
          XM_008769295.1 XP_008767517.1 14 - 15 5209 62
          GenBank mRNA BC168999.1 - 11 - 12 1702 62

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of receptor internalization
          biological_process
          cerebellar Purkinje cell differentiation
          cytoplasmic mRNA processing body assembly
          stress granule assembly
          negative regulation of multicellular organism growth
          neuron projection morphogenesis
          homeostasis of number of cells
          neuromuscular process
          molecular_function
          epidermal growth factor receptor binding
          protein C-terminus binding
          poly(A) RNA binding
          DBD domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P108948 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P108948 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P108948 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P108948 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P389098 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P373389 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10762088 - -
          Affymetrix Rat Gene 1.0 ST Array 10762105 - -
          Affymetrix Rat Gene 1.0 ST Array 10762104 - -
          Affymetrix Rat Gene 1.0 ST Array 10762096 - -
          Affymetrix Rat Gene 1.0 ST Array 10762095 - -
          Affymetrix Rat Gene 1.0 ST Array 10762102 - -
          Affymetrix Rat Gene 1.0 ST Array 10762089 - -
          Affymetrix Rat Gene 1.0 ST Array 10762106 - -
          Affymetrix Rat Gene 1.0 ST Array 10762098 - -
          Affymetrix Rat Gene 1.0 ST Array 10762091 - -
          Affymetrix Rat Gene 1.0 ST Array 10762100 - -
          Affymetrix Rat Gene 1.0 ST Array 10762099 - -
          Affymetrix Rat Gene 1.0 ST Array 10762101 - -
          Affymetrix Rat Gene 1.0 ST Array 10762094 - -
          Affymetrix Rat Gene 1.0 ST Array 10762092 - -
          Affymetrix Rat Gene 1.0 ST Array 10762093 - -
          Affymetrix Rat Gene 1.0 ST Array 10762103 - -
          Affymetrix Rat Gene 1.0 ST Array 10762097 - -
          Affymetrix Rat Gene 1.0 ST Array 10762086 - -
          Affymetrix Rat Gene 1.1 ST Array 10762085 - -
          Rat Genome 230 2.0 Array 1391158_a_at - -
          Rat Genome 230 2.0 Array 1382480_PM_at - -
          Rat Genome 230 2.0 Array 1391158_PM_a_at - -
          Rat Genome 230 2.0 Array 1382480_at - -
          Affymetrix HT Rat Focus Array 1391158_a_at - -
          Affymetrix HT Rat Focus Array 1391158_PM_a_at - -
          Rat Expression Set 230 1382480_PM_at - -
          Rat Expression Set 230 1391158_a_at - -
          Rat Expression Set 230 1382480_at - -
          Rat Expression Set 230 1391158_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10762085 - -
          Rat Genome U34 Set rc_AI011816_at - -
          Rat Genome U34 Set rc_AA899901_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22392212 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371229 - -
          RatRef-12 Expression BeadChip ILMN_53770 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline