Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01499782_m1
          See other ELN GE Assays ›
          Gene Symbol:
          Eln
          Gene Name:
          elastin
          Gene Aliases:
          RATTREL11, TREL11, Trela, Trela26
          Chromosome Location:
          Chr.12: 24978478 - 25021864 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01499782_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eln
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012722.1 NP_036854.1 35 - 36 2536 95
          GenBank mRNA BC085910.1 - 35 - 36 2536 7097661 95
          J04035.1 - 4 - 5 158 95
          M60647.1 - 35 - 36 2498 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000439.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eln

          Panther Classification:

          Molecular Function -

          structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel remodeling
          skeletal muscle tissue development
          aging
          extracellular matrix organization
          regulation of actin filament polymerization
          aorta development
          response to diuretic
          stress fiber assembly
          cellular response to fibroblast growth factor stimulus
          elastic fiber assembly
          response to hyperoxia
          cellular response to copper ion
          cellular response to L-ascorbic acid
          cellular response to retinoic acid
          cellular response to organic cyclic compound
          cellular response to hypoxia
          cellular response to dexamethasone stimulus
          cellular response to transforming growth factor beta stimulus
          extracellular matrix structural constituent
          extracellular matrix constituent conferring elasticity
          extracellular matrix binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P140374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768180 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75333 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114214 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13227 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53968 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161697 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138919 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15663 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15663 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138919 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15663 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P138919 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P454386 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37382 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40023 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103758 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43082 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10757740 - -
          Affymetrix Rat Gene 1.0 ST Array 10757741 - -
          Affymetrix Rat Gene 1.0 ST Array 10757735 - -
          Affymetrix Rat Gene 1.0 ST Array 10757757 - -
          Affymetrix Rat Gene 1.0 ST Array 10757765 - -
          Affymetrix Rat Gene 1.0 ST Array 10757734 - -
          Affymetrix Rat Gene 1.0 ST Array 10757761 - -
          Affymetrix Rat Gene 1.0 ST Array 10757752 - -
          Affymetrix Rat Gene 1.0 ST Array 10757762 - -
          Affymetrix Rat Gene 1.0 ST Array 10757730 - -
          Affymetrix Rat Gene 1.0 ST Array 10757743 - -
          Affymetrix Rat Gene 1.0 ST Array 10757753 - -
          Affymetrix Rat Gene 1.0 ST Array 10757739 - -
          Affymetrix Rat Gene 1.0 ST Array 10757732 - -
          Affymetrix Rat Gene 1.0 ST Array 10757751 - -
          Affymetrix Rat Gene 1.0 ST Array 10757748 - -
          Affymetrix Rat Gene 1.0 ST Array 10757767 - -
          Affymetrix Rat Gene 1.0 ST Array 10757731 - -
          Affymetrix Rat Gene 1.0 ST Array 10757738 - -
          Affymetrix Rat Gene 1.0 ST Array 10757728 - -
          Affymetrix Rat Gene 1.0 ST Array 10757733 - -
          Affymetrix Rat Gene 1.0 ST Array 10757758 - -
          Affymetrix Rat Gene 1.0 ST Array 10757737 - -
          Affymetrix Rat Gene 1.0 ST Array 10757760 - -
          Affymetrix Rat Gene 1.0 ST Array 10757736 - -
          Affymetrix Rat Gene 1.0 ST Array 10757727 - -
          Affymetrix Rat Gene 1.0 ST Array 10757763 - -
          Affymetrix Rat Gene 1.0 ST Array 10757744 - -
          Affymetrix Rat Gene 1.0 ST Array 10757754 - -
          Affymetrix Rat Gene 1.0 ST Array 10757766 - -
          Affymetrix Rat Gene 1.0 ST Array 10757745 - -
          Affymetrix Rat Gene 1.0 ST Array 10757747 - -
          Affymetrix Rat Gene 1.0 ST Array 10757742 - -
          Affymetrix Rat Gene 1.0 ST Array 10757749 - -
          Affymetrix Rat Gene 1.0 ST Array 10757764 - -
          Affymetrix Rat Gene 1.0 ST Array 10757746 - -
          Affymetrix Rat Gene 1.0 ST Array 10757755 - -
          Affymetrix Rat Gene 1.0 ST Array 10757759 - -
          Affymetrix Rat Gene 1.0 ST Array 10757729 - -
          Affymetrix Rat Gene 1.0 ST Array 10757750 - -
          Affymetrix Rat Gene 1.1 ST Array 10757726 - -
          Rat Genome 230 2.0 Array 1388111_PM_at - -
          Rat Genome 230 2.0 Array 1388111_at - -
          Rat Genome 230 2.0 Array 1395753_at - -
          Rat Genome 230 2.0 Array 1395753_PM_at - -
          Affymetrix HT Rat Focus Array 1388111_PM_at - -
          Affymetrix HT Rat Focus Array 1395753_at - -
          Affymetrix HT Rat Focus Array 1388111_at - -
          Affymetrix HT Rat Focus Array 1395753_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200003909.rn.1 TC1200000439.rn.1 RTA TC
          Rat Expression Set 230 1388111_at - -
          Rat Expression Set 230 1388111_PM_at - -
          Rat Expression Set 230 1395753_at - -
          Rat Expression Set 230 1395753_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10757726 - -
          Rat Genome U34 Set J04035_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20718514 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358233 - -
          RatRef-12 Expression BeadChip ILMN_53896 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline