Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01500022_m1
          See other TMEM248 GE Assays ›
          Gene Symbol:
          Tmem248
          Gene Name:
          transmembrane protein 248
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 29931728 - 29958050 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01500022_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem248
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004204.2 NP_001004204.1 8 - 9 1339 89
          XM_006249230.2 XP_006249292.1 5 - 6 1041 89
          XM_006249231.2 XP_006249293.1 5 - 6 962 89
          GenBank mRNA BC079161.1 - 5 - 6 1013 7129411 89
          BC090318.1 - 8 - 9 1339 7113430 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001429.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem248

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1022403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274293 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88430 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82395 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22759 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1022403 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1022403 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1022403 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1022403 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71810 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81956 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14125 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10761334 - -
          Affymetrix Rat Gene 1.0 ST Array 10761330 - -
          Affymetrix Rat Gene 1.0 ST Array 10761337 - -
          Affymetrix Rat Gene 1.0 ST Array 10761332 - -
          Affymetrix Rat Gene 1.0 ST Array 10761331 - -
          Affymetrix Rat Gene 1.0 ST Array 10761336 - -
          Affymetrix Rat Gene 1.0 ST Array 10761338 - -
          Affymetrix Rat Gene 1.0 ST Array 10761335 - -
          Affymetrix Rat Gene 1.0 ST Array 10761333 - -
          Affymetrix Rat Gene 1.0 ST Array 10761339 - -
          Affymetrix Rat Gene 1.0 ST Array 10761340 - -
          Affymetrix Rat Gene 1.1 ST Array 10761329 - -
          Rat Genome 230 2.0 Array 1371905_at - -
          Rat Genome 230 2.0 Array 1371905_PM_at - -
          Affymetrix HT Rat Focus Array 1371905_PM_at - -
          Affymetrix HT Rat Focus Array 1371905_at - -
          Affymetrix Rat Clariom S Assay PSR1200012057.rn.1 TC1200001429.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012056.rn.1 TC1200001429.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012058.rn.1 TC1200001429.rn.1 RTA TC
          Rat Expression Set 230 1371905_PM_at - -
          Rat Expression Set 230 1371905_at - -
          Affymetrix Rat Gene 1.0 ST Array 10761329 - -
          Rat Genome U34 Set rc_AI012932_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358419 - -
          RatRef-12 Expression BeadChip ILMN_47918 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline