Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01500325_m1
          See other CEP41 GE Assays ›
          Gene Symbol:
          Cep41
          Gene Name:
          centrosomal protein 41
          Gene Aliases:
          Tsga14
          Chromosome Location:
          Chr.4: 57966783 - 58006963 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01500325_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cep41
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025770.2 NP_001020941.1 8 - 9 942 69
          XM_006236229.2 XP_006236291.1 10 - 11 1117 69
          GenBank mRNA BC098832.1 - 8 - 9 944 7378587 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002611.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cep41

          Gene Ontology Categories:

          Function(s)

          protein transport
          protein polyglutamylation
          cilium assembly

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P945759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568435 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79016 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150647 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P945759 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P945759 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P945759 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P945759 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103673 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77568 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10861596 - -
          Affymetrix Rat Gene 1.0 ST Array 10861593 - -
          Affymetrix Rat Gene 1.0 ST Array 10861602 - -
          Affymetrix Rat Gene 1.0 ST Array 10861600 - -
          Affymetrix Rat Gene 1.0 ST Array 10861597 - -
          Affymetrix Rat Gene 1.0 ST Array 10861601 - -
          Affymetrix Rat Gene 1.0 ST Array 10861592 - -
          Affymetrix Rat Gene 1.0 ST Array 10861594 - -
          Affymetrix Rat Gene 1.0 ST Array 10861595 - -
          Affymetrix Rat Gene 1.0 ST Array 10861598 - -
          Affymetrix Rat Gene 1.0 ST Array 10861599 - -
          Affymetrix Rat Gene 1.1 ST Array 10861591 - -
          Rat Genome 230 2.0 Array 1382409_PM_at - -
          Rat Genome 230 2.0 Array 1382409_at - -
          Rat Genome 230 2.0 Array 1382865_at - -
          Rat Genome 230 2.0 Array 1382865_PM_at - -
          Affymetrix HT Rat Focus Array 1382865_PM_at - -
          Affymetrix HT Rat Focus Array 1382865_at - -
          Affymetrix HT Rat Focus Array 1382409_at - -
          Affymetrix HT Rat Focus Array 1382409_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400018542.rn.1 TC0400002611.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400018543.rn.1 TC0400002611.rn.1 RTA TC
          Rat Expression Set 230 1382865_at - -
          Rat Expression Set 230 1382865_PM_at - -
          Rat Expression Set 230 1382409_PM_at - -
          Rat Expression Set 230 1382409_at - -
          Affymetrix Rat Gene 1.0 ST Array 10861591 - -
          Rat Genome U34 Set rc_AI072515_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20951790 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372140 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline