Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01500585_m1
          See other CPA2 GE Assays ›
          Gene Symbol:
          Cpa2
          Gene Name:
          carboxypeptidase A2 (pancreatic)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 57855416 - 57879239 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01500585_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cpa2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013083.2 NP_001013101.2 10 - 11 1071 66
          GenBank mRNA CF249156.1 - 2 - 3 168 66
          CF249511.1 - 4 - 5 435 66
          FQ226028.1 - 7 - 8 754 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000549.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cpa2

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          carboxypeptidase activity
          metallocarboxypeptidase activity
          protein binding
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1039213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905565 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54639 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101774 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1039213 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1039213 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1039213 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1039213 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34936 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85563 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40698 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10854301 - -
          Affymetrix Rat Gene 1.0 ST Array 10854297 - -
          Affymetrix Rat Gene 1.0 ST Array 10854302 - -
          Affymetrix Rat Gene 1.0 ST Array 10854300 - -
          Affymetrix Rat Gene 1.0 ST Array 10854295 - -
          Affymetrix Rat Gene 1.0 ST Array 10854294 - -
          Affymetrix Rat Gene 1.0 ST Array 10854299 - -
          Affymetrix Rat Gene 1.0 ST Array 10854296 - -
          Affymetrix Rat Gene 1.0 ST Array 10854298 - -
          Affymetrix Rat Gene 1.0 ST Array 10854293 - -
          Affymetrix Rat Gene 1.0 ST Array 10854292 - -
          Affymetrix Rat Gene 1.1 ST Array 10854291 - -
          Rat Genome 230 2.0 Array 1393397_PM_at - -
          Rat Genome 230 2.0 Array 1393397_at - -
          Affymetrix HT Rat Focus Array 1393397_at - -
          Affymetrix HT Rat Focus Array 1393397_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400003939.rn.1 TC0400000549.rn.1 RTA TC
          Rat Expression Set 230 1393397_at - -
          Rat Expression Set 230 1393397_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10854291 - -
          Rat Genome U34 Set M23721_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20858952 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650345 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline