Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01501298_m1
          See other KLHDC10 GE Assays ›
          Gene Symbol:
          Klhdc10
          Gene Name:
          kelch domain containing 10
          Gene Aliases:
          RGD1309792
          Chromosome Location:
          Chr.4: 57715979 - 57773800 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01501298_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klhdc10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017456.1 NP_001017456.1 8 - 9 1099 154
          XM_006236201.2 XP_006236263.1 9 - 10 1255 154
          GenBank mRNA BC085351.1 - 8 - 9 1099 7115168 154

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000546.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klhdc10

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P126366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282566 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19459 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99114 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50753 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7505 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39569 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148261 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32578 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P126366 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P126366 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P126366 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P126366 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87773 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72790 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23818 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65191 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104490 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10854274 - -
          Affymetrix Rat Gene 1.0 ST Array 10854280 - -
          Affymetrix Rat Gene 1.0 ST Array 10854273 - -
          Affymetrix Rat Gene 1.0 ST Array 10854278 - -
          Affymetrix Rat Gene 1.0 ST Array 10854282 - -
          Affymetrix Rat Gene 1.0 ST Array 10854281 - -
          Affymetrix Rat Gene 1.0 ST Array 10854276 - -
          Affymetrix Rat Gene 1.0 ST Array 10854283 - -
          Affymetrix Rat Gene 1.0 ST Array 10854279 - -
          Affymetrix Rat Gene 1.0 ST Array 10854277 - -
          Affymetrix Rat Gene 1.1 ST Array 10854272 - -
          Affymetrix Rat Clariom S Assay PSR0400003899.rn.1 TC0400000546.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10854272 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21320169 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362375 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline