Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01501359_m1
          See other ADAMTS19 GE Assays ›
          Gene Symbol:
          Adamts19
          Gene Name:
          ADAM metallopeptidase with thrombospondin type 1 motif, 19
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 53915807 - 54099856 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01501359_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adamts19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108433.1 NP_001101903.1 5 - 6 1677 66
          XM_006254755.2 XP_006254817.1 5 - 6 1672 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000587.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Protease 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          integrin-mediated signaling pathway
          metalloendopeptidase activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P045764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93318 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60331 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117793 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145030 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73420 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36736 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16822 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81833 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70762 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22382 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147851 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61891 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171181 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68994 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133162 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19251 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38188 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63791 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33254 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167477 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155808 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159835 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73358 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122219 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P045764 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P045764 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P045764 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P550840 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P102291 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60068 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35981 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24491 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38228 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87272 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10801944 - -
          Affymetrix Rat Gene 1.0 ST Array 10801959 - -
          Affymetrix Rat Gene 1.0 ST Array 10801936 - -
          Affymetrix Rat Gene 1.0 ST Array 10801949 - -
          Affymetrix Rat Gene 1.0 ST Array 10801957 - -
          Affymetrix Rat Gene 1.0 ST Array 10801951 - -
          Affymetrix Rat Gene 1.0 ST Array 10801945 - -
          Affymetrix Rat Gene 1.0 ST Array 10801954 - -
          Affymetrix Rat Gene 1.0 ST Array 10801938 - -
          Affymetrix Rat Gene 1.0 ST Array 10801941 - -
          Affymetrix Rat Gene 1.0 ST Array 10801956 - -
          Affymetrix Rat Gene 1.0 ST Array 10801953 - -
          Affymetrix Rat Gene 1.0 ST Array 10801937 - -
          Affymetrix Rat Gene 1.0 ST Array 10801958 - -
          Affymetrix Rat Gene 1.0 ST Array 10801943 - -
          Affymetrix Rat Gene 1.0 ST Array 10801950 - -
          Affymetrix Rat Gene 1.0 ST Array 10801947 - -
          Affymetrix Rat Gene 1.0 ST Array 10801952 - -
          Affymetrix Rat Gene 1.0 ST Array 10801946 - -
          Affymetrix Rat Gene 1.0 ST Array 10801948 - -
          Affymetrix Rat Gene 1.0 ST Array 10801955 - -
          Affymetrix Rat Gene 1.0 ST Array 10801942 - -
          Affymetrix Rat Gene 1.0 ST Array 10801939 - -
          Affymetrix Rat Gene 1.0 ST Array 10801940 - -
          Affymetrix Rat Gene 1.1 ST Array 10801935 - -
          Rat Genome 230 2.0 Array 1379586_PM_at - -
          Rat Genome 230 2.0 Array 1379586_at - -
          Affymetrix Rat Clariom S Assay PSR1800004280.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004267.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004268.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004272.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004279.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004283.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004284.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004286.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004273.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004266.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004270.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004276.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004288.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004289.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004269.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004271.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004277.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004278.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004281.rn.1 TC1800000587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004282.rn.1 TC1800000587.rn.1 RTA TC
          Rat Expression Set 230 1379586_PM_at - -
          Rat Expression Set 230 1379586_at - -
          Affymetrix Rat Gene 1.0 ST Array 10801935 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21131281 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362126 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline