Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01501410_m1
          See other SIRT3 GE Assays ›
          Gene Symbol:
          Sirt3
          Gene Name:
          sirtuin 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 213613502 - 213636194 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01501410_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sirt3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106313.2 NP_001099783.1 6 - 7 925 76
          XM_006230526.2 XP_006230588.1 6 - 7 1005 76
          XM_006230529.2 XP_006230591.1 6 - 7 1619 76
          XM_008759979.1 XP_008758201.1 6 - 7 1045 76
          XM_008759980.1 XP_008758202.1 5 - 6 883 76
          XM_008759981.1 XP_008758203.1 6 - 7 913 76
          GenBank mRNA BC167006.1 - 6 - 7 925 7314283 76
          EU886468.1 - 6 - 7 951 76
          FQ213357.1 - 5 - 6 751 76
          FQ218084.1 - 6 - 7 956 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006911.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein deacetylation
          aging
          aerobic respiration
          positive regulation of insulin secretion
          peptidyl-lysine deacetylation
          negative regulation of ERK1 and ERK2 cascade
          histone H3 deacetylation
          negative regulation of reactive oxygen species metabolic process
          negative regulation of peptidyl-lysine acetylation
          zinc ion binding
          enzyme binding
          NAD-dependent histone deacetylase activity (H3-K14 specific)
          NAD+ binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P089946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246987 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65092 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5388 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99375 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P089946 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P089946 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P089946 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P534782 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57148 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85727 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10726666 - -
          Affymetrix Rat Gene 1.0 ST Array 10726668 - -
          Affymetrix Rat Gene 1.0 ST Array 10726664 - -
          Affymetrix Rat Gene 1.0 ST Array 10726663 - -
          Affymetrix Rat Gene 1.0 ST Array 10726662 - -
          Affymetrix Rat Gene 1.0 ST Array 10726665 - -
          Affymetrix Rat Gene 1.1 ST Array 10726661 - -
          Rat Genome 230 2.0 Array 1372925_PM_at - -
          Rat Genome 230 2.0 Array 1372925_at - -
          Affymetrix HT Rat Focus Array 1372925_PM_at - -
          Affymetrix HT Rat Focus Array 1372925_at - -
          Affymetrix Rat Clariom S Assay PSR0100054409.rn.1 TC0100006911.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100054408.rn.1 TC0100006911.rn.1 RTA TC
          Rat Expression Set 230 1372925_PM_at - -
          Rat Expression Set 230 1372925_at - -
          Affymetrix Rat Gene 1.0 ST Array 10726661 - -
          Rat Genome U34 Set rc_AI233135_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21260312 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354765 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline