Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01501483_m1
          See other TJP2 GE Assays ›
          Gene Symbol:
          Tjp2
          Gene Name:
          tight junction protein 2
          Gene Aliases:
          ZO-2
          Chromosome Location:
          Chr.1: 241945816 - 242084042 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01501483_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tjp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053773.1 NP_446225.1 22 - 23 3555 110
          XM_006231175.2 XP_006231237.1 22 - 23 4637 110
          XM_006231176.2 XP_006231238.1 21 - 22 4527 110
          XM_006231177.2 XP_006231239.1 21 - 22 3868 110
          XM_006231178.2 XP_006231240.1 21 - 22 6797 110
          XM_006231179.2 XP_006231241.1 21 - 22 3701 110
          XM_006231181.2 XP_006231243.1 22 - 23 3656 110
          XM_006231182.2 XP_006231244.1 20 - 21 3757 110
          XM_008760302.1 XP_008758524.1 22 - 23 3397 110
          GenBank mRNA BC103479.1 - 22 - 23 3555 7936785 110
          U75916.1 - 15 - 16 2188 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007436.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tjp2

          Panther Classification:

          Molecular Function -

          tight junction

          Gene Ontology Categories:

          Function(s) Process(es)

          response to organic substance
          intestinal absorption
          establishment of endothelial intestinal barrier
          regulation of membrane permeability
          protein C-terminus binding
          protein domain specific binding
          protein binding, bridging

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P484836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811203 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132628 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112651 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46799 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99210 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51318 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168660 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62535 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36433 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97715 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105407 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64107 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33675 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60785 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P484836 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P484836 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P484836 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P484836 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700369452 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98294 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 888 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97915 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92819 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41695 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33203 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51852 - -
          Agilent Rat cDNA Microarray (G4105A) 700369452 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10729411 - -
          Affymetrix Rat Gene 1.0 ST Array 10729419 - -
          Affymetrix Rat Gene 1.0 ST Array 10729424 - -
          Affymetrix Rat Gene 1.0 ST Array 10729413 - -
          Affymetrix Rat Gene 1.0 ST Array 10729423 - -
          Affymetrix Rat Gene 1.0 ST Array 10729421 - -
          Affymetrix Rat Gene 1.0 ST Array 10729410 - -
          Affymetrix Rat Gene 1.0 ST Array 10729429 - -
          Affymetrix Rat Gene 1.0 ST Array 10729431 - -
          Affymetrix Rat Gene 1.0 ST Array 10729437 - -
          Affymetrix Rat Gene 1.0 ST Array 10729414 - -
          Affymetrix Rat Gene 1.0 ST Array 10729415 - -
          Affymetrix Rat Gene 1.0 ST Array 10729430 - -
          Affymetrix Rat Gene 1.0 ST Array 10729434 - -
          Affymetrix Rat Gene 1.0 ST Array 10729432 - -
          Affymetrix Rat Gene 1.0 ST Array 10729420 - -
          Affymetrix Rat Gene 1.0 ST Array 10729418 - -
          Affymetrix Rat Gene 1.0 ST Array 10729425 - -
          Affymetrix Rat Gene 1.0 ST Array 10729433 - -
          Affymetrix Rat Gene 1.0 ST Array 10729422 - -
          Affymetrix Rat Gene 1.0 ST Array 10729428 - -
          Affymetrix Rat Gene 1.0 ST Array 10729426 - -
          Affymetrix Rat Gene 1.0 ST Array 10729412 - -
          Affymetrix Rat Gene 1.0 ST Array 10729427 - -
          Affymetrix Rat Gene 1.1 ST Array 10729409 - -
          Rat Genome 230 2.0 Array 1370940_at - -
          Rat Genome 230 2.0 Array 1370940_PM_at - -
          Affymetrix HT Rat Focus Array 1370940_PM_at - -
          Affymetrix HT Rat Focus Array 1370940_at - -
          Affymetrix Rat Clariom S Assay PSR0100060033.rn.1 TC0100007436.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100060035.rn.1 TC0100007436.rn.1 RTA TC
          Rat Expression Set 230 1370940_at - -
          Rat Expression Set 230 1370940_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10729409 - -
          Rat Genome U34 Set U75916_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21611264 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366515 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline