Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01501559_g1
          See other PSAT1 GE Assays ›
          Gene Symbol:
          Psat1
          Gene Name:
          phosphoserine aminotransferase 1
          Gene Aliases:
          Psa1
          Chromosome Location:
          Chr.1: 233124089 - 233145941 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01501559_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psat1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198738.2 NP_942033.2 7 - 8 889 104
          GenBank mRNA AY239016.1 - 7 - 8 877 104
          BC079352.1 - 7 - 8 889 7108218 104

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psat1

          Panther Classification:

          Molecular Function -

          transaminase

          Gene Ontology Categories:

          Function(s) Process(es)

          L-serine biosynthetic process
          O-phospho-L-serine:2-oxoglutarate aminotransferase activity
          pyridoxal phosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P162599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766234 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52619 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 255 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99047 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P162599 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P162599 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P162599 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P149161 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89807 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94809 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36120 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10729101 - -
          Affymetrix Rat Gene 1.0 ST Array 10729099 - -
          Affymetrix Rat Gene 1.0 ST Array 10729098 - -
          Affymetrix Rat Gene 1.0 ST Array 10823612 - -
          Affymetrix Rat Gene 1.0 ST Array 10729097 - -
          Affymetrix Rat Gene 1.0 ST Array 10729100 - -
          Affymetrix Rat Gene 1.1 ST Array 10729096 - -
          Affymetrix Rat Gene 1.1 ST Array 10823611 - -
          Rat Genome 230 2.0 Array 1372665_at - -
          Rat Genome 230 2.0 Array 1372665_PM_at - -
          Affymetrix HT Rat Focus Array 1372665_PM_at - -
          Affymetrix HT Rat Focus Array 1372665_at - -
          Affymetrix Rat Clariom S Assay PSR0100059345.rn.1 TC0100007349.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100059349.rn.1 TC0100007349.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100059344.rn.1 TC0100007349.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100059348.rn.1 TC0100007349.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100059347.rn.1 TC0100007349.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100059346.rn.1 TC0100007349.rn.1 RTA TC
          Rat Expression Set 230 1372665_at - -
          Rat Expression Set 230 1372665_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10729096 - -
          Affymetrix Rat Gene 1.0 ST Array 10823611 - -
          Rat Genome U34 Set rc_AI102868_at - -
          Rat Genome U34 Set rc_AA850568_at - -
          Rat Genome U34 Set rc_AI230228_at - -
          Rat Genome U34 Set rc_AI102868_g_at - -
          Rat Genome U34 Set rc_AI102868_at - -
          Rat Genome U34 Set rc_AI102868_g_at - -
          Rat Genome U34 Set rc_AA850568_at - -
          Rat Genome U34 Set rc_AI230228_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21584564 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline