Search
Search

Cdc37l1| Gene Symbol: Cdc37l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011941.1 | NP_001011941.1 | 5 - 6 | 836 | 143 | |
| XM_006231201.2 | XP_006231263.1 | 6 - 7 | 995 | 143 | ||
| XM_006231202.2 | XP_006231264.1 | 6 - 7 | 995 | 143 | ||
| XM_008760322.1 | XP_008758544.1 | 6 - 7 | 995 | 143 | ||
| GenBank mRNA | BC083761.1 | - | 5 - 6 | 836 | 7192975 | 143 |
Set Membership: |
|