Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01502070_m1
          See other MAP3K11 GE Assays ›
          Gene Symbol:
          Map3k11
          Gene Name:
          mitogen-activated protein kinase kinase kinase 11
          Gene Aliases:
          Mlk-3, Mlk3, RHOE
          Chromosome Location:
          Chr.1: 221043119 - 221056420 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01502070_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Map3k11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013150.1 NP_001013168.1 8 - 9 2272 149
          GenBank mRNA BC081952.1 - 8 - 9 2272 7122664 149

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003067.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Map3k11

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          protein phosphorylation
          microtubule-based process
          JNK cascade
          activation of JNKK activity
          activation of JUN kinase activity
          cell death
          cell proliferation
          peptidyl-tyrosine phosphorylation
          positive regulation of apoptotic process
          positive regulation of JUN kinase activity
          positive regulation of neuron apoptotic process
          positive regulation of JNK cascade
          protein autophosphorylation
          protein serine/threonine kinase activity
          JUN kinase kinase kinase activity
          MAP kinase kinase kinase activity
          protein tyrosine kinase activity
          protein binding
          ATP binding
          mitogen-activated protein kinase kinase binding
          mitogen-activated protein kinase kinase kinase binding
          identical protein binding
          protein homodimerization activity
          Rac GTPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P224840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95925 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29915 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P224840 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P224840 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P224840 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P261243 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31800 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10713114 - -
          Affymetrix Rat Gene 1.0 ST Array 10713115 - -
          Affymetrix Rat Gene 1.0 ST Array 10713106 - -
          Affymetrix Rat Gene 1.0 ST Array 10713112 - -
          Affymetrix Rat Gene 1.0 ST Array 10713108 - -
          Affymetrix Rat Gene 1.0 ST Array 10713113 - -
          Affymetrix Rat Gene 1.0 ST Array 10713105 - -
          Affymetrix Rat Gene 1.0 ST Array 10713103 - -
          Affymetrix Rat Gene 1.0 ST Array 10713107 - -
          Affymetrix Rat Gene 1.0 ST Array 10713110 - -
          Affymetrix Rat Gene 1.0 ST Array 10713111 - -
          Affymetrix Rat Gene 1.0 ST Array 10713104 - -
          Affymetrix Rat Gene 1.0 ST Array 10713109 - -
          Affymetrix Rat Gene 1.1 ST Array 10713102 - -
          Rat Genome 230 2.0 Array 1374997_at - -
          Rat Genome 230 2.0 Array 1374997_PM_at - -
          Affymetrix HT Rat Focus Array 1374997_PM_at - -
          Affymetrix HT Rat Focus Array 1374997_at - -
          Affymetrix Rat Clariom S Assay PSR0100024903.rn.1 TC0100003067.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100024905.rn.1 TC0100003067.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100024904.rn.1 TC0100003067.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100024902.rn.1 TC0100003067.rn.1 RTA TC
          Rat Expression Set 230 1374997_at - -
          Rat Expression Set 230 1374997_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10713102 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21208015 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650004 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline