Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01502357_m1
          See other COL14A1 GE Assays ›
          Gene Symbol:
          Col14a1
          Gene Name:
          collagen, type XIV, alpha 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 95054877 - 95274073 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01502357_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Col14a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001130548.1 NP_001124020.1 8 - 9 1150 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001128.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          extracellular matrix structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ventricular cardiac muscle tissue development
          collagen fibril organization
          homeostasis of number of cells within a tissue
          regulation of cell growth involved in cardiac muscle cell development
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P093194 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P093189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56910 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70795 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60360 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60226 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114839 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19320 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156594 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27930 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19766 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42804 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122497 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127072 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150160 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24074 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70858 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173214 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154076 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45691 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80375 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98379 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37663 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63891 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61572 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49007 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35821 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21510 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48171 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093189 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093194 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093194 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093189 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P093189 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P093194 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P171271 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18681 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10896596 - -
          Affymetrix Rat Gene 1.0 ST Array 10896595 - -
          Affymetrix Rat Gene 1.0 ST Array 10896588 - -
          Affymetrix Rat Gene 1.0 ST Array 10896572 - -
          Affymetrix Rat Gene 1.0 ST Array 10896584 - -
          Affymetrix Rat Gene 1.0 ST Array 10896566 - -
          Affymetrix Rat Gene 1.0 ST Array 10896564 - -
          Affymetrix Rat Gene 1.0 ST Array 10896586 - -
          Affymetrix Rat Gene 1.0 ST Array 10896585 - -
          Affymetrix Rat Gene 1.0 ST Array 10896565 - -
          Affymetrix Rat Gene 1.0 ST Array 10896562 - -
          Affymetrix Rat Gene 1.0 ST Array 10896599 - -
          Affymetrix Rat Gene 1.0 ST Array 10896598 - -
          Affymetrix Rat Gene 1.0 ST Array 10896597 - -
          Affymetrix Rat Gene 1.0 ST Array 10896574 - -
          Affymetrix Rat Gene 1.0 ST Array 10896571 - -
          Affymetrix Rat Gene 1.0 ST Array 10896561 - -
          Affymetrix Rat Gene 1.0 ST Array 10896570 - -
          Affymetrix Rat Gene 1.0 ST Array 10896593 - -
          Affymetrix Rat Gene 1.0 ST Array 10896587 - -
          Affymetrix Rat Gene 1.0 ST Array 10896591 - -
          Affymetrix Rat Gene 1.0 ST Array 10896563 - -
          Affymetrix Rat Gene 1.0 ST Array 10896583 - -
          Affymetrix Rat Gene 1.0 ST Array 10896575 - -
          Affymetrix Rat Gene 1.0 ST Array 10896582 - -
          Affymetrix Rat Gene 1.0 ST Array 10896559 - -
          Affymetrix Rat Gene 1.0 ST Array 10896604 - -
          Affymetrix Rat Gene 1.0 ST Array 10896590 - -
          Affymetrix Rat Gene 1.0 ST Array 10896589 - -
          Affymetrix Rat Gene 1.0 ST Array 10896567 - -
          Affymetrix Rat Gene 1.0 ST Array 10896603 - -
          Affymetrix Rat Gene 1.0 ST Array 10896581 - -
          Affymetrix Rat Gene 1.0 ST Array 10896600 - -
          Affymetrix Rat Gene 1.0 ST Array 10896560 - -
          Affymetrix Rat Gene 1.0 ST Array 10896579 - -
          Affymetrix Rat Gene 1.0 ST Array 10896578 - -
          Affymetrix Rat Gene 1.0 ST Array 10896580 - -
          Affymetrix Rat Gene 1.0 ST Array 10896569 - -
          Affymetrix Rat Gene 1.0 ST Array 10896573 - -
          Affymetrix Rat Gene 1.0 ST Array 10896577 - -
          Affymetrix Rat Gene 1.0 ST Array 10896592 - -
          Affymetrix Rat Gene 1.0 ST Array 10896594 - -
          Affymetrix Rat Gene 1.0 ST Array 10896576 - -
          Affymetrix Rat Gene 1.0 ST Array 10896568 - -
          Affymetrix Rat Gene 1.0 ST Array 10896602 - -
          Affymetrix Rat Gene 1.0 ST Array 10896601 - -
          Affymetrix Rat Gene 1.1 ST Array 10896558 - -
          Rat Genome 230 2.0 Array 1376105_PM_at - -
          Rat Genome 230 2.0 Array 1376170_at - -
          Rat Genome 230 2.0 Array 1376170_PM_at - -
          Rat Genome 230 2.0 Array 1376105_at - -
          Affymetrix HT Rat Focus Array 1376170_PM_at - -
          Affymetrix HT Rat Focus Array 1376105_PM_at - -
          Affymetrix HT Rat Focus Array 1376170_at - -
          Affymetrix HT Rat Focus Array 1376105_at - -
          Affymetrix Rat Clariom S Assay PSR0700008243.rn.1 TC0700001128.rn.1 RTA TC
          Rat Expression Set 230 1376105_at - -
          Rat Expression Set 230 1376105_PM_at - -
          Rat Expression Set 230 1376170_at - -
          Rat Expression Set 230 1376170_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10896558 - -
          Rat Genome U34 Set rc_AA998128_at - -
          Rat Genome U34 Set rc_AI060224_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20859593 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374380 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline