Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01502638_m1
          See other SREBF2 GE Assays ›
          Gene Symbol:
          Srebf2
          Gene Name:
          sterol regulatory element binding transcription factor 2
          Gene Aliases:
          SREBP-2, SREBP2, Srebf2_retired
          Chromosome Location:
          Chr.7: 123381082 - 123438605 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01502638_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Srebf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033694.1 NP_001028866.1 17 - 18 3212 61
          XM_006242079.2 XP_006242141.1 17 - 18 3567 61
          GenBank mRNA BC101902.1 - 17 - 18 3212 7934137 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001520.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Srebf2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          spermatogenesis
          aging
          cholesterol metabolic process
          lipid biosynthetic process
          response to hormone
          response to lead ion
          positive regulation of cholesterol storage
          response to lipid
          response to drug
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          response to low-density lipoprotein particle
          regulation of lipid transport by negative regulation of transcription from RNA polymerase II promoter
          negative regulation of cholesterol efflux
          regulation of cholesterol homeostasis
          C-8 sterol isomerase activity
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein C-terminus binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein dimerization activity
          E-box binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P092209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147691 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162855 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126503 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173180 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108625 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141639 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092209 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092209 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P092209 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P382747 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P504215 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12857 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61677 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72855 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98687 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28946 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49777 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100726 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10898107 - -
          Affymetrix Rat Gene 1.0 ST Array 10898105 - -
          Affymetrix Rat Gene 1.0 ST Array 10898116 - -
          Affymetrix Rat Gene 1.0 ST Array 10898094 - -
          Affymetrix Rat Gene 1.0 ST Array 10898098 - -
          Affymetrix Rat Gene 1.0 ST Array 10898096 - -
          Affymetrix Rat Gene 1.0 ST Array 10898113 - -
          Affymetrix Rat Gene 1.0 ST Array 10898101 - -
          Affymetrix Rat Gene 1.0 ST Array 10898100 - -
          Affymetrix Rat Gene 1.0 ST Array 10898097 - -
          Affymetrix Rat Gene 1.0 ST Array 10898108 - -
          Affymetrix Rat Gene 1.0 ST Array 10898114 - -
          Affymetrix Rat Gene 1.0 ST Array 10898109 - -
          Affymetrix Rat Gene 1.0 ST Array 10898111 - -
          Affymetrix Rat Gene 1.0 ST Array 10898092 - -
          Affymetrix Rat Gene 1.0 ST Array 10898110 - -
          Affymetrix Rat Gene 1.0 ST Array 10898115 - -
          Affymetrix Rat Gene 1.0 ST Array 10898099 - -
          Affymetrix Rat Gene 1.0 ST Array 10898102 - -
          Affymetrix Rat Gene 1.0 ST Array 10898104 - -
          Affymetrix Rat Gene 1.0 ST Array 10898112 - -
          Affymetrix Rat Gene 1.0 ST Array 10898095 - -
          Affymetrix Rat Gene 1.0 ST Array 10898093 - -
          Affymetrix Rat Gene 1.0 ST Array 10898106 - -
          Affymetrix Rat Gene 1.1 ST Array 10898091 - -
          Rat Genome 230 2.0 Array 1377489_at - -
          Rat Genome 230 2.0 Array 1371979_PM_at - -
          Rat Genome 230 2.0 Array 1377489_PM_at - -
          Rat Genome 230 2.0 Array 1371979_at - -
          Affymetrix HT Rat Focus Array 1377489_at - -
          Affymetrix HT Rat Focus Array 1371979_at - -
          Affymetrix HT Rat Focus Array 1371979_PM_at - -
          Affymetrix HT Rat Focus Array 1377489_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700011651.rn.1 TC0700001520.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10898091 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22160023 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354176 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline